Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD104 Rat anti-Mouse, BB515, Clone: 346-11A, BD

Rat Monoclonal Antibody

Manufacturer:  BD Biosciences 565600

Catalog No. BDB565600


Add to cart

Description

Description

The 346-11A monoclonal antibody specifically recognizes an epitope at the beginning of the cysteine-rich repeat region of the 200-kDa integrin β4 chain (CD104), which is found on the cell surface as a heterodimeric complex with the integrin α6 chain (CD49f). The α6β4 (CD49f/CD104) complex binds to laminins and is expressed on the basal surface of a variety of epithelial cell types, particularly on stratified squamous epithelia, and is also found in peripheral nerves, in certain subsets of endothelial cells, and on immature thymocytes. It has also been identified on a number of tumor tissues and participates in tumor progression events. L ocalization of both human and mouse epidermal α6β4 integrin to hemidesmosomes suggests that this heterodimer plays a role in epidermal adhesion to the basement membrane.

Specifications

Specifications

CD104
346-11A
BB515
Aqueous buffered solution containing ≤0.09% sodium azide.
Rat
IgG2a, κ
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
0.2mg/mL
β4 integrin; Integrin β4 chain; Integrin beta-4; Itgb4; ITB4
Affinity Purified
Tumor-associated antigen TSP-180 from a BALB/c mouse lung carcinoma
Affinity Chromatography
RUO
Primary
Murine
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit