Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD155 Rat anti-Mouse, PE, Clone: 3F1, BD

Rat Monoclonal Antibody

Manufacturer:  BD Biosciences 565797

Catalog No. BDB565797


Add to cart

Description

Description

The 3F1 monoclonal antibody specifically binds to CD155, which is also known as Poliovirus receptor (Pvr) or Tumor-associated antigen 1 (Taa1). CD155 is a type I transmembrane glycoprotein that belongs to the Ig supergene family. CD155 is an adhesion receptor that binds to different ligands including nectin-3, CD96, CD226, TIGIT, and the extracellular matrix protein vitronectin. It is highly expressed on double positive thymocytes and variably expressed on mature thymocytes and T cells, including regulatory T cells and NKT cells. CD155 is also differentially expressed on subsets of B cells, plasma cells, dendritic cells, and monocytes. CD155 expression is upregulated by activated T cells, B cells, and dendritic cells. CD155 is involved in forming adherens junctions between adjacent epithelial or endothelial cells. CD155 plays roles in regulating cell growth, adhesion, motility, migration, and cell-mediated cytotoxicity. CD155-deficient mice exhibit impaired secondary humoral immune responses to orally administered antigens.

Specifications

Specifications

CD155
3F1
PE
Aqueous buffered solution containing ≤0.09% sodium azide.
Rat
IgG2a, κ
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
0.2mg/mL
Poliovirus receptor; Pvr; Tumor-associated antigen 1; Taa1; Tage4; HVED
Affinity Purified
Mouse CD155 Recombinant Protein
Affinity Chromatography
RUO
Primary
Murine
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit