Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat anti-CD43/Sialophorin, Clone: R2/60, Biotin, Novus Biologicals™
Rat Monoclonal Antibody
$364.00 - $728.00
Specifications
Antigen | CD43/Sialophorin |
---|---|
Clone | R2/60 |
Concentration | 0.5 mg/mL |
Dilution | Flow Cytometry 1:10-1:1000 |
Applications | Flow Cytometry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP14364705
|
Novus Biologicals
NBP1436470.05MG |
0.05 mg |
Each for $364.00
|
|
|||||
NBP14364701
|
Novus Biologicals
NBP1436470.1MG |
0.1 mg |
Each for $582.00
|
|
|||||
NBP1436475
|
Novus Biologicals
NBP1436470.5MG |
0.5 mg |
Each for $728.00
|
|
|||||
Description
CD43/Sialophorin Monoclonal specifically detects CD43/Sialophorin in Mouse samples. It is validated for Flow Cytometry.Specifications
CD43/Sialophorin | |
0.5 mg/mL | |
Flow Cytometry | |
Biotin | |
RUO | |
Mouse | |
CD43 antigen, CD43), Galactoglycoprotein, GALGP, Leukocyte sialoglycoprotein, Sialophorin, sialophorin (gpL115, leukosialin, CD43) | |
SPN | |
IgM | |
Affinity Purified |
R2/60 | |
Flow Cytometry 1:10-1:1000 | |
Monoclonal | |
Rat | |
Immunology | |
Aqueous buffer, may contain carrier protein/stabilizer (proprietary) with 0.09% Sodium Azide | |
6693 | |
The immunogen for this antibody was CD43. | |
Primary | |
Store at 4C. Do not freeze. |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title