Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat anti-CD63, Biotin, Clone: 7G4.2E8, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236567B

Catalog No. NBP236567B


Add to cart

Description

Description

CD63 Monoclonal antibody specifically detects CD63 in Human, Mouse samples. It is validated for Western Blot.
Specifications

Specifications

CD63
7G4.2E8
Western Blot
P41731
CD63 antigen, CD63 antigen (melanoma 1 antigen), CD63 molecule, Granulophysin, LAMP-3, Lysosomal-associated membrane protein 3, ME491, melanoma 1 antigen, Melanoma-associated antigen ME491, melanoma-associated antigen MLA1, MLA1lysosome-associated membrane glycoprotein 3, Ocular melanoma-associated antigen, OMA81H, Tetraspanin-30, tspan-30, TSPAN30granulophysin
Rat
IgG2a Lambda
0.1 ml
Autophagy, Cytokine Research
Primary
967
Western Blot
Biotin
PBS with 0.05% Sodium Azide
Purified
CD63
Partial recombinant mouse CD63 protein (between amino acids 100-250) [UniProt P41731]
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit