Learn More
Rat anti-CD63, DyLight 488, Clone: 7G4.2E8, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236567G
Catalog No. NBP236567G
Add to cart
Description
Description
CD63 Monoclonal antibody specifically detects CD63 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Flow (Intracellular).
Specifications
Specifications
CD63
7G4.2E8
Western Blot, Flow Cytometry, Flow (Intracellular)
P41731
CD63 antigen, CD63 antigen (melanoma 1 antigen), CD63 molecule, Granulophysin, LAMP-3, Lysosomal-associated membrane protein 3, ME491, melanoma 1 antigen, Melanoma-associated antigen ME491, melanoma-associated antigen MLA1, MLA1lysosome-associated membrane glycoprotein 3, Ocular melanoma-associated antigen, OMA81H, Tetraspanin-30, tspan-30, TSPAN30granulophysin
Rat
IgG2a Lambda
0.1 ml
Autophagy, Cytokine Research
Primary
967
Flow Cytometry, Western Blot
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
Purified
CD63
Partial recombinant mouse CD63 protein (between amino acids 100-250) [UniProt P41731]
Protein G purified
RUO
Store at 4C in the dark.
Monoclonal
Human, Mouse