Learn More
Rat anti-CD90.2, Clone: 53-2.1, Biotin, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP1436090.1MG
 View more versions of this product
Catalog No. NBP14360901
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
CD90.2 Monoclonal antibody specifically detects CD90.2 in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
CD90.2
53-2.1
Biotin
Aqueous buffer, may contain carrier protein/stabilizer (proprietary) with 0.09% Sodium Azide
CD90, CD90 antigen, CDw90, FLJ33325, Thy-1 antigen, Thy-1 cell surface antigen, thy-1 membrane glycoprotein, Thy-1 T-cell antigen
Rat
IgG2a Kappa
0.1 mg
Hematopoietic Stem Cell Markers, Immunology, Innate Immunity, Mesenchymal Stem Cell Markers, Signal Transduction, Stem Cell Markers
Primary
7070
Flow Cytometry
0.5 mg/mL
Flow Cytometry 1:10-1:1000
Affinity Purified
THY1
The immunogen for this antibody was CD90.2.
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
Mouse