Learn More
Rat anti-DC-SIGN/CD209, Clone: LWC06, FITC, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP1439650.025MG
 View more versions of this product
Catalog No. NBP14396503
Add to cart
Description
Description
DC-SIGN/CD209 Monoclonal antibody specifically detects DC-SIGN/CD209 in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
DC-SIGN/CD209
LWC06
FITC
Aqueous buffer, may contain carrier protein/stabilizer (proprietary) with 0.09% Sodium Azide
CD209 antigendendritic cell-specific intracellular adhesion molecules (ICAM)-3 grabbingnon-integrin, CD209 molecule, CDSIGNHIV gpl20-binding protein, CLEC4LC-type lectin domain family 4 member L, DC-SIGN1C-type lectin domain family 4, member L, DC-SIGNMGC129965, Dendritic cell-specific ICAM-3-grabbing non-integrin 1
Rat
IgG2a
0.025 mg
Adaptive Immunity, Immunology, Virology Bacteria and Parasites
Primary
30835
Flow Cytometry
0.5 mg/mL
Flow Cytometry 1:10-1:1000
Affinity Purified
CD209
The immunogen for this antibody was CIRE.
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
Mouse