Learn More
Rat anti-Dendritic Cell Marker, Clone: 33D1, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP1432840.5MG
 View more versions of this product
Catalog No. NBP1432845
Add to cart
Description
Description
Dendritic Cell Marker Monoclonal antibody specifically detects Dendritic Cell Marker in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen.
Specifications
Specifications
Dendritic Cell Marker
33D1
Unconjugated
PBS, pH 7.2 with 0.09% Sodium Azide
DCs, Dendritic cell
The immunogen for this antibody was Dendritic Cell Marker.
Protein A or G purified
RUO
Primary
Mouse
Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen)
0.5 mg/mL
Flow Cytometry 1:10-1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Affinity Purified
Rat
IgG2b Kappa
0.5 mg
Store at 4C. Do not freeze.
Monoclonal