Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat anti-Dendritic Cell Marker, Clone: 33D1, PE, Novus Biologicals™
Rat Monoclonal Antibody
$582.00 - $642.00
Specifications
Antigen | Dendritic Cell Marker |
---|---|
Clone | 33D1 |
Concentration | 0.2 mg/mL |
Dilution | Flow Cytometry 1:10-1:1000 |
Applications | Flow Cytometry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP20015310
|
Novus Biologicals
NBP200153100UG |
100 μg |
Each for $582.00
|
|
|||||
NBP20015326
|
Novus Biologicals
NBP200153200UG |
200 μg |
Each for $642.00
|
|
|||||
Description
Dendritic Cell Marker Monoclonal specifically detects Dendritic Cell Marker in Mouse samples. It is validated for Flow Cytometry.Specifications
Dendritic Cell Marker | |
0.2 mg/mL | |
Flow Cytometry | |
PE | |
RUO | |
Aqueous buffer, may contain carrier protein/stabilizer (proprietary) with 0.09% Sodium Azide | |
The immunogen for this antibody was Dendritic Cell Marker. | |
Primary | |
Store at 4C. Do not freeze. |
33D1 | |
Flow Cytometry 1:10-1:1000 | |
Monoclonal | |
Rat | |
Mouse | |
DCs, Dendritic cell | |
IgG2b κ | |
Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title