Learn More
Rat anti-Flt-3/Flk-2/CD135, Clone: A2F10, PE-Cy5, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP1430780.1MG
 View more versions of this product
Catalog No. NBP14307801
Add to cart
Description
Description
Flt-3/Flk-2/CD135 Monoclonal antibody specifically detects Flt-3/Flk-2/CD135 in Human, Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
Flt-3/Flk-2/CD135
A2F10
Flow Cytometry 1:10-1:1000
Affinity Purified
FLT3
The immunogen for this antibody was CD135.
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
Mouse
Flow Cytometry
PE-Cyanine5
Aqueous buffer, may contain carrier protein/stabilizer (proprietary) with 0.09% Sodium Azide
CD135, CD135 antigen, EC 2.7.10, fetal liver kinase 2, FL cytokine receptor, FLK2, FLT-3, FLT3 receptor tyrosine kinase, Fms-like tyrosine kinase 3, fms-related tyrosine kinase 3, growth factor receptor tyrosine kinase type III, Stem cell tyrosine kinase 1, STK-1, STK1EC 2.7.10.1, Tyrosine-protein kinase receptor FLT3
Rat
IgG2a Kappa
0.1 mg
Cell Cycle and Replication
Primary
2322