Learn More
Galectin-3 Rat anti-Mouse, Clone: M3/38, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP1433130.5MG
 View more versions of this product
Catalog No. NBP1433135
Add to cart
Description
Description
Galectin-3 Monoclonal specifically detects Galectin-3 in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular).
Specifications
Specifications
Galectin-3
M3/38
Unconjugated
Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide
Carbohydrate-binding protein 35, CBP 35,35 kDa lectin, Gal-3, galactin-3, Galactose-specific lectin 3, Galactoside-binding protein, GALBPCBP35, galectin-3, GALIG, IgE-binding protein, L31, L-31, Laminin-binding protein, Lectin L-29, lectin, galactoside-binding, soluble, 3, LGALS2, MAC-2, Mac-2 antigen, MAC2GAL3
Rat
IgG2a Kappa
0.5 mg
Apoptosis, Cancer, Cellular Markers, Tumor Suppressors
Primary
3958
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
0.5 mg/mL
Western Blot 1:100-1:2000, Flow Cytometry 1:10-1:1000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Flow (Intracellular)
Affinity Purified
LGALS3
The immunogen for this antibody was Galectin3 Mac2.
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
Mouse