Learn More
Gata3, Rat, Clone: KT63, Abnova™
Rat monoclonal antibody raised against Gata3.
Manufacturer:  Abnova Corporation MAB10148
Catalog No. 89-108-403
Add to cart
Description
Description
Specifications
Specifications
Gata3
KT63
Rat monoclonal antibody raised against Gata3.
In PBS (0.09% sodium azide)
Liquid
Gata3
Mouse Gata3.
Protein G purification
RUO
Primary
14462
Immunofluorescence, Western Blot
Unconjugated
Western Blot (0.1-1 ug/mL) Immunofluorescence (1-10 ug/mL) The optimal working dilution should be determined by the end user.
Gata3
Gata-3
Rat
IgG2a
100 ug
Store at 4°C for one month. For long term storage store at -20°C or -80°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Mouse