Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat anti-GDF-15, Clone: 6D12.H10.E4, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP244214

Catalog No. NBP244214


Add to cart

Description

Description

Ensure accurate, reproducible results in Western Blot, ELISA

GDF-15 Monoclonal antibody specifically detects GDF-15 in Human, Mouse samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

GDF-15
6D12.H10.E4
Unconjugated
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
growth differentiation factor 15, growth/differentiation factor 15, Macrophage inhibitory cytokine 1, MIC-1NSAID-activated gene 1 protein, MIC1Prostate differentiation factor, NAG-1NSAID-regulated gene 1 protein, NSAID (nonsteroidal inflammatory drug)-activated protein 1, PDFGDF-15, PLABNRG-1, Placental bone morphogenetic protein, Placental TGF-beta, PTGFBPTGF-beta
Rat
IgG2a Lambda
0.1 mg
Hepatic Stem Cell Markers, Stem Cell Markers
Primary
9518
Western Blot, ELISA
1 mg/mL
Western Blot 1:1000, ELISA 1:2000
Purified
GDF15
This Protein-A purified antibody was prepared by repeated immunizations with an MBP-tagged recombinant protein produced in E.coli corresponding to full-length mouse GDF-15 protein. Cross reactivity to MBP was removed via cross-adsorption chromatography.
Protein G purified
RUO
Store at -20°C. Avoid freeze-thaw cycles.
Monoclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only