Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat anti-GM-CSF, Clone: SPM469, Azide Free, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP2347740.1 MG

 View more versions of this product

Catalog No. NBP23477401


Add to cart

Description

Description

GM-CSF Monoclonal antibody specifically detects GM-CSF in Human, Primate samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Frozen, CyTOF-ready, Block/Neutralize.
Specifications

Specifications

GM-CSF
SPM469
Unconjugated
10mM PBS - BSA free with No Preservative
Purified
CSF2
Recombinant human GM-CSF protein
22 kDa
0.1 mg
Cytokine Research, Immunology
Primary
1437
Protein A purified
ELISA, Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen), Immunoprecipitation, Western Blot
1.0 mg/mL
Western Blot 0.5-1ug/ml, Flow Cytometry 0.5-1ug/million cells, ELISA 1-5ug/ml for coating, Immunocytochemistry/Immunofluorescence 0.5-1ug/ml, Immunoprecipitation 0.5-1ug/500ug protein lysate, Immunohistochemistry-Frozen 0.5-1ug/ml, Blocking/Neutralizing, CyTOF-ready
P04141
colony stimulating factor 2 (granulocyte-macrophage), Colony-stimulating factor, CSF, GM-CSF, GMCSFgranulocyte-macrophage colony-stimulating factor, granulocyte-macrophage colony stimulating factor, MGC131935, MGC138897, molgramostin, sargramostim
Rat
IgG2a Kappa
Protein A purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Monoclonal
Granulocyte/macrophage - Colony-stimulating factor (GM-CSF) is a hematopoietic factor that is produced by activated T-cells, B-cells, mast cells, macrophages, fibroblasts, and endothelial cells. In addition to supporting colony formation of granulocyte/macrophage progenitors, GM-CSF is a growth factor for erythroid, megakaryocyte, and eosinophil progenitors.
Human, Primate
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only