Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat anti-gp96/HSP90B1/GRP94, Clone: HSP90B1/1192, Novus Biologicals™

Rat Monoclonal Antibody

$129.00 - $299.00

Specifications

Antigen gp96/HSP90B1/GRP94
Clone HSP90B1/1192
Concentration 0.2mg/mL
Immunogen Recombinant full-length human HSP90B1 protein
Gene ID (Entrez) 7184
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP24469100 View Documents Novus BiologicalsSupplier Diversity Partner
NBP2446910.02MG
0.02 mg Each for $129.00
Add to cart
 
NBP24469101 View Documents Novus BiologicalsSupplier Diversity Partner
NBP2446910.1MG
0.1 mg Each for $299.00
Add to cart
 
NBP24469102 View Documents Novus BiologicalsSupplier Diversity Partner
NBP2446910.2MG
0.2 mg This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

Ensure accurate, reproducible results in Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence

gp96/HSP90B1/GRP94 Monoclonal antibody specifically detects gp96/HSP90B1/GRP94 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry-Paraffin, Immunofluorescence.
Specifications

Specifications

gp96/HSP90B1/GRP94
0.2mg/mL
7184
Monoclonal
Rat
IgG2a Kappa
Purified
RUO
Protein A or G purified
HSP90B1/1192
Recombinant full-length human HSP90B1 protein
10mM PBS and 0.05% BSA with 0.05% Sodium Azide
Primary
Recognizes a protein of 94kDa, which is identified as the glucose-regulated protein 94 (grp94) and also tumor rejection antigen (gp96). Grp94 shows a high degree of sequence homology with the heat shock protein 90 (hsp90). This MAb is highly specific to grp94 and shows minimal cross-reaction with other members of the HSP90 family. Grp s are a class of proteins unresponsive to heat shock and are induced by glucose deprivation. Grp94 has been briefly studied as a prognostic factor in breast cancer.
Unconjugated
Western Blot, Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence
Store at 4C.

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit