Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD294 (CRTH2) Rat anti-Human, BUV395, Clone: BM16, BD Optibuild™

Rat Monoclonal Antibody

Manufacturer:  BD Biosciences 740319

Catalog No. BDB740319


Add to cart

Description

Description

The BM16 monoclonal antibody specifically binds to CD294. CD294 is encoded by PTGDR2 (Prostaglandin D2 receptor 2) and is also known as CRTH2 (chemoattractant receptor-homologous molecule expressed on Th2 cells), GPR44 (G protein-coupled receptor 44), DL1R, and DP2. CD294 is a member of the G protein-coupled leukocyte chemoattractant receptor family. CD294 is expressed on Th2 cells and type-2 innate lymphoid cells (ILC2), but not Th1 type cells. CD294 is detectable on CD4+ T cells in fresh PBMC but not on B cells and NK cells. CD294 is also expressed on peripheral blood basophils and eosinophils, suggesting its involvement allergic reactions. Phenotypic analysis of CD4+ T cells expressing CRTH2 demonstrated that they were also CD45RA-negative and CD45RO+ and CD25+. These cells produce Th2- but little or no Th1-type cytokines upon stimulation with PMA and Ionomycin.

The antibody was conjugated to BD Horizon™ BUV395 which is part of the BD Horizon Brilliant™ Ultraviolet family of dyes. This dye has been exclusively developed by BD Biosciences to have minimal spillover into other detectors, making it an optimal choice for multicolor flow cytometry. With an Ex Max at 348nm and an Em Max at 395nm, BD Horizon BUV395 can be excited with a 355nm laser and detected with a 379/28 filter.

Specifications

Specifications

CD294 (CRTH2)
BM16
Brilliant Ultraviolet 395
Aqueous buffered solution containing ≤0.09% sodium azide.
Rat
IgG2a, κ
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
0.2mg/mL
CRTH2; PTGDR2 ; Prostaglandin D2 receptor 2; DL1R; DP2; GPR44
Affinity Purified
Human CRTH2 Transfected Cell Line
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.
RUO
Primary
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only