Learn More
Rat anti-ICOS, Clone: 7E.17G9, Novus Biologicals™
Manufacturer:  Novus Biologicals NBP1431290.05MG
Catalog No. NBP14312905
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
ICOS
7E.17G9
Unconjugated
Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide
Activation-inducible lymphocyte immunomediatory molecule, AILIMCD278, CD278 antigen, CVID1, inducible costimulator, inducible T-cell costimulator, inducible T-cell co-stimulator, MGC39850
Rat
IgG2b Kappa
0.05 mg
Store at 4C. Do not freeze.
Monoclonal
Mouse
Flow Cytometry
0.2 mg/mL
Flow Cytometry 1:10-1:1000
Affinity Purified
ICOS
The immunogen for this antibody was ICOS.
Protein A or G purified
RUO
Primary
29851