Learn More
Rat anti-ICOS, Clone: 7E.17G9, PE-Cy5, Novus Biologicals™
Manufacturer:  Novus Biologicals NBP1430660.05MG
 View more versions of this product
Catalog No. NBP14306605
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
ICOS
7E.17G9
Flow Cytometry 1:10-1:1000
Affinity Purified
ICOS
The immunogen for this antibody was ICOS.
Protein A or G purified
RUO
Primary
29851
Flow Cytometry
PE-Cyanine5
Aqueous buffer, may contain carrier protein/stabilizer (proprietary) with 0.09% Sodium Azide
Activation-inducible lymphocyte immunomediatory molecule, AILIMCD278, CD278 antigen, CVID1, inducible costimulator, inducible T-cell costimulator, inducible T-cell co-stimulator, MGC39850
Rat
IgG2b Kappa
0.05 mg
Store at 4C. Do not freeze.
Monoclonal
Mouse