Learn More
Rat anti-IgG1, κ, Isotype Control, Brilliant Blue 515, Clone: R3-34, BD
Manufacturer:  BD Biosciences 564610
Catalog No. BDB564610
Add to cart
Description
Description
Specifications
Specifications
IgG1 κ
R3-34
Brilliant Blue 515
Affinity Purified
Mouse immunoglobulin
Affinity Chromatography
RUO
Monoclonal
Flow Cytometry
0.2mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
Rat
IgG1, κ
50μg
Store undiluted at 4°C. Do not freeze. Protect from light.
Not Provided