Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat anti-IgG2c,k, Brilliant Ultraviolet 737, Clone: A23-1, BD

Manufacturer:  BD Biosciences 565524

Catalog No. BDB565524


Add to cart

Specifications

Specifications

IgG2c,k
A23-1
Brilliant Ultraviolet 737
Affinity Purified
TNP-keyhole limpet hemocyanin
Affinity Chromatography
RUO
Primary
Not Provided
Flow Cytometry
0.2mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
Rat
LOU
50μg
Store undiluted at 4°C. Do not freeze. Protect from light.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit