Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rat anti-IL-13, DyLight 405, Clone: 13A, Novus Biologicals™

Rat Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP143239V

Catalog No. NBP143239V


Add to cart

Description

Description

Ensure accurate, reproducible results in ELISA

IL-13 Monoclonal antibody specifically detects IL-13 in Mouse samples. It is validated for Flow Cytometry, ELISA.
Specifications

Specifications

IL-13
13A
DyLight 405
50mM Sodium Borate with 0.05% Sodium Azide
ALRHMGC116789, BHR1interleukin-13, IL-13MGC116788, interleukin 13, MGC116786, NC30, P600
Rat
IgG1 Kappa
0.1 ml
Asthma, Cancer, Cytokine Research, Immunology, Innate Immunity
Primary
3596
ELISA, Flow Cytometry
0.5mg/mL
Flow Cytometry, ELISA
Affinity Purified
IL13
The immunogen for this antibody was IL13.
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only