Learn More
IL-2 R beta Rat anti-Mouse, Clone: TM-b1, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP1433550.5MG
 View more versions of this product
Catalog No. NBP1433555
Add to cart
Description
Description
IL-2 R beta Monoclonal antibody specifically detects IL-2 R beta in Mouse samples. It is validated for Flow Cytometry, Immunoprecipitation, Block/Neutralize.
Specifications
Specifications
IL-2 R beta
TM-b1
Unconjugated
Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide
CD122, CD122 antigen, high affinity IL-2 receptor beta subunit, High affinity IL-2 receptor subunit beta, IL-2 receptor subunit beta, IL-2R subunit beta, IL-2RB, interleukin 15 receptor, beta, interleukin 2 receptor, beta, interleukin-2 receptor subunit beta, P70-75, p75
Rat
IgG2b
0.5 mg
Store at 4C. Do not freeze.
Monoclonal
Mouse
Flow Cytometry, Immunoprecipitation, Blocking Assay
0.5 mg/mL
Flow Cytometry 1:10-1:1000, Immunoprecipitation 1:10-1:500, Blocking/Neutralizing 50-200 molar excess
Affinity Purified
IL2RB
The immunogen for this antibody was Purified CD122.
Protein A or G purified
RUO
Primary
3560