Learn More
Rat anti-Integrin alpha 4 beta 7/LPAM-1, Clone: DATK32, Novus Biologicals™
Rat Monoclonal Antibody
Manufacturer:  Novus Biologicals NBP1432830.05MG
 View more versions of this product
Catalog No. NBP14328305
Add to cart
Description
Description
Integrin alpha 4 beta 7/LPAM-1 Monoclonal antibody specifically detects Integrin alpha 4 beta 7/LPAM-1 in Mouse samples. It is validated for Flow Cytometry.
Specifications
Specifications
Integrin alpha 4 beta 7/LPAM-1
DATK32
Unconjugated
Aqueous with 0.09% Sodium Azide
Affinity Purified
Rat
IgG2a Kappa
0.05 mg
Innate Immunity
Primary
3676
Flow Cytometry, Functional Assay
0.5 mg/mL
Flow Cytometry 1:10-1:1000, Functional
P13612, Q00651, P26011
269C wild type, CD49 antigen-like family member D, CD49d, CD49d antigen, CD49Dantigen CD49D, alpha-4 subunit of VLA-4 receptor, IA4, integrin alpha 4, integrin alpha-4, integrin alpha-4 subunit, Integrin alpha-IV, integrin, alpha 4 (antigen CD49D, alpha 4 subunit of VLA-4 receptor), MGC90518, very late activation protein 4 receptor, alpha 4 subunit, VLA-4 subunit alpha
The immunogen for this antibody was LPAM1.
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
Mouse