Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Laminin gamma 1 Antibody (A5), Novus Biologicals™
Rat Monoclonal Antibody has been used in 3 publications
$265.00 - $506.50
Specifications
Antigen | Laminin gamma 1 |
---|---|
Clone | A5 |
Concentration | 0.2mg/mL |
Dilution | Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, Immunofluorescence 1 - 2 ug/ml |
Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP24475100
|
Novus Biologicals
NBP2447510.02MG |
0.02 mg |
Each for $265.00
|
|
|||||
NBP24475101
|
Novus Biologicals
NBP2447510.1MG |
0.1 mg |
Each for $506.50
|
|
|||||
NBP24475102
|
Novus Biologicals
NBP2447510.2MG |
0.2 mg | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
Ensure accurate, reproducible results in Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence
Laminin gamma 1 Monoclonal specifically detects Laminin gamma 1 in Human, Mouse, Bovine samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunofluorescence.Specifications
Laminin gamma 1 | |
0.2mg/mL | |
Flow Cytometry, Immunohistochemistry (Paraffin), Immunofluorescence | |
Unconjugated | |
Rat | |
Apoptosis, Cancer, Cytoskeleton Markers, Extracellular Matrix, Tumor Suppressors | |
10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
LAMB2Laminin-7 subunit gamma, Laminin B2 chain, laminin subunit gamma-1, laminin, gamma 1 (formerly LAMB2), Laminin-1 subunit gamma, Laminin-10 subunit gamma, Laminin-11 subunit gamma, Laminin-2 subunit gamma, Laminin-3 subunit gamma, Laminin-4 subunit gamma, Laminin-6 subunit gamma, Laminin-8 subunit gamma, Laminin-9 subunit gamma, MGC87297, S-LAM gamma, S-laminin subunit gamma | |
LAMC1 | |
IgG2a κ | |
Protein A or G purified | |
Laminins are large hetero-trimeric, non-collagenous glycoproteins composed of alpha, beta, and gamma chains. This MAb reacts with laminin B2/1 chain of ∼210kDa and does not cross-react with other basement membrane components or fibronectin. Its specificity was established by immunoprecipitation and immunofluorescence of human skeletal muscle and kidney with laminin chain-specific MAbs. Epithelial sheets in vivo are separated from the mesenchymal elements of the stroma by a thin layer of a specialized type of extracellular matrix termed the basement membrane (BM). This structure consists of individual components, some of which are ubiquitous in BMs and some are not. The ubiquitous ones comprise laminin (LN), entactin/nidogen (EN), collagen type IV (CIV), and large heparan sulfate proteoglycan (HSPG), which interact specifically with each other to form a continuous and regular BM. Alterations of BM integrity, from local discontinuities up to complete loss, are described in many types |
A5 | |
Flow Cytometry 0.5 - 1 ug/million cells in 0.1 ml, Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, Immunofluorescence 1 - 2 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human, Mouse | |
Q5VYE7, Q6NVY8 | |
3915 | |
Murine EHS laminin preparation | |
Primary | |
Store at 4C. | |
210 kDa |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title