Learn More
M-CSFR/CD115 Rat anti-Mouse, Clone: AFS98, Novus Biologicals™
Rat Monoclonal Antibody has been used in 2 publications
Manufacturer:  Novus Biologicals NBP1433630.05MG
 View more versions of this product
Catalog No. NBP14336305
Add to cart
Description
Description
M-CSF R/CD115 Monoclonal antibody specifically detects M-CSF R/CD115 in Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry-Frozen, Block/Neutralize.
Specifications
Specifications
M-CSF R/CD115
AFS98
Unconjugated
Aqueous, may contain carrier protein/stabilizer. with 0.09% Sodium Azide
Affinity Purified
CSF1R
The immunogen for this antibody was CD115.
Protein A or G purified
RUO
Store at 4C. Do not freeze.
Monoclonal
Mouse
Western Blot, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Blocking Assay
0.5 mg/mL
Western Blot 1:100-1:2000, Flow Cytometry < 1 ug/10^6 cells, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500, Blocking/Neutralizing
P07333
CD115, CD115 antigen, C-FMS, colony stimulating factor 1 receptor, CSF-1-R, CSFR, EC 2.7.10.1, FMS proto-oncogene, FMSFIM2, macrophage colony stimulating factor I receptor, macrophage colony-stimulating factor 1 receptor, McDonough feline sarcoma viral (v-fms) oncogene homolog, Proto-oncogene c-Fms
Rat
IgG2a Kappa
0.05 mg
Myeloid derived Suppressor Cell, Protein Kinase
Primary
1436