Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Rat anti-MHC Class II, Clone: M5/114.15.2, FITC, Novus Biologicals™
Rat Monoclonal Antibody
$364.00 - $946.00
Specifications
Antigen | MHC Class II |
---|---|
Clone | M5/114.15.2 |
Concentration | 0.5 mg/mL |
Dilution | Flow Cytometry 1:10-1:1000 |
Applications | Flow Cytometry |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP14395005
|
Novus Biologicals
NBP1439500.05MG |
0.05 mg |
Each for $364.00
|
|
|||||
NBP1439505
|
Novus Biologicals
NBP1439500.5MG |
0.5 mg |
Each for $946.00
|
|
|||||
Description
MHC class II (I-A/I-E) Monoclonal specifically detects MHC class II (I-A/I-E) in Human, Mouse samples. It is validated for Flow Cytometry.Specifications
MHC Class II | |
0.5 mg/mL | |
Flow Cytometry | |
FITC | |
RUO | |
Mouse | |
class II histocompatibility antigen, M alpha chain, D6S222E, DMA, HLA class II histocompatibility antigen, DM alpha chain, major histocompatibility complex, class II, DM alpha, MHC class II antigen DMA, Really interesting new gene 6 protein, RING6HLADM | |
HLA-DMA | |
IgG2b κ | |
Affinity Purified | |
The monoclonal antibody reacts with the mouse major histocompatibility complex class II, both I-A and I-E subregion-encoded glycoproteins (I-A b, I-A d, I-A q, I-E d, I-E k, not I-A f, I-A k, or I-A s). It detects a polymorphic determinant present on B cells, monocytes, macrophages, dendritic cells, and activated T lymphocytes from mice carrying the H-2 b, H-2 d, H-2 q, H-2 p, H-2 r and H-2 u but not from mice carrying the H-2 s or H-2 f haplotypes. The M5/114 mAb is reported to inhibit I-A-restricted T cell responses of the H-2 b, H-2 d, H-2 q, H-2 u but not H-2 f, H-2 k, or H-2 s haplotypes. |
M5/114.15.2 | |
Flow Cytometry 1:10-1:1000 | |
Monoclonal | |
Rat | |
Adaptive Immunity, Immune System Diseases, Immunology, Major Histocompatibility Complexes, Transcription Factors and Regulators | |
Aqueous buffer, may contain carrier protein/stabilizer (proprietary) with 0.09% Sodium Azide | |
3108 | |
The immunogen for this antibody was MHC Class II. | |
Primary | |
Store at 4C. Do not freeze. |
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title