Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD105 Rat anti-Mouse, Brilliant Violet 711, Clone: MJ7/18, BD Optibuild™

Rat Monoclonal Antibody

Manufacturer:  BD Biosciences 740808

Catalog No. BDB740808


Add to cart

Description

Description

The MJ7/18 monoclonal antibody specifically binds to mouse CD105 (also known as endoglin) which is a homodimer of 90-kDa subunits and is predominantly expressed on vascular endothelial cells. High levels of mouse endoglin mRNA have been reported to be detectable in the ovary, uterus, NCTC-2071 fibroblasts, and to a lesser extent, in heart, muscle and stromal cells in connective tissue of various organs. Endoglin has been reported to play an essential role in embryonic angiogenesis. Both mouse and human endoglin display strong amino-acid sequence homology to the transmembrane and cytoplasmic regions of the type III TGF-ß receptor.

The antibody was conjugated to BD Horizon™ BV711 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 711-nm. BD Horizon BV711 can be excited by the violet laser and detected in a filter used to detect Cy™5.5 / Alexa Fluor™ 700-like dyes (eg, 712/20-nm filter). Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy5.5 detectors. However, the spillover can be corrected through compensation as with any other dye combination.

Specifications

Specifications

CD105
MJ7/18
Brilliant Violet 711
Aqueous buffered solution containing ≤0.09% sodium azide.
Rat
IgG2a, κ
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
0.2mg/mL
Endoglin; Edg; EGLN; Eng; MJ7/18 antigen; S-endoglin
Affinity Purified
Mouse skin (inflamed)
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.
RUO
Primary
Murine
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only