Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD106 Rat anti-Mouse, Brilliant Violet 711, Clone: 429 (MVCAM.A), BD Optibuild™

Rat Monoclonal Antibody

Manufacturer:  BD Biosciences 740675

Catalog No. BDB740675


Add to cart

Description

Description

The 429 monoclonal antibody specifically binds to both the long (~110 kDa) transmembrane-spanning form and the truncated (~47 kDa) GPI-linked form of vascular cell adhesion molecule-1 (VCAM-1, CD106). CD106 is constitutively expressed on bone marrow stromal cells, myeloid cells, and splenic dendritic cells. Its expression on endothelial cells is upregulated by inflammatory cytokines and in certain pathologic conditions. CD106 expression has also been detected on apoptotic thymocytes, splenocytes, and lymphoid cell lines. VCAM-1 is a counter-receptor for VLA-4 ( α 4β 1 integrin) and LPAM-1 ( α 4β 7 integrin), and the 429 antibody partially blocks VCAM-1-mediated binding functions. Source of the immunogen was the mouse preadipose cell line PA6.

Specifications

Specifications

CD106
429 (MVCAM.A)
Brilliant Violet 711
Aqueous buffered solution containing ≤0.09% sodium azide.
Rat
IgG2a, κ
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
0.2mg/mL
Vcam-1; Vascular cell adhesion molecule 1; Vascular cell adhesion protein 1
Affinity Purified
Mouse preadipose cell line PA6
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.
RUO
Primary
Murine
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only