Learn More
CD106 Rat anti-Mouse, Brilliant Violet 786, Clone: 429 (MVCAM.A), BD Optibuild™
Rat Monoclonal Antibody
Supplier: BD Biosciences 740865
Description
The 429 monoclonal antibody specifically binds to both the long (∽110 kDa) transmembrane-spanning form and the truncated (∽47 kDa) GPI-linked form of vascular cell adhesion molecule-1 (VCAM-1, CD106). CD106 is constitutively expressed on bone marrow stromal cells, myeloid cells, and splenic dendritic cells. Its expression on endothelial cells is upregulated by inflammatory cytokines and in certain pathologic conditions. CD106 expression has also been detected on apoptotic thymocytes, splenocytes, and lymphoid cell lines. VCAM-1 is a counter-receptor for VLA-4 ( α 4β 1 integrin) and LPAM-1 ( α 4β 7 integrin), and the 429 antibody partially blocks VCAM-1-mediated binding functions. Source of the immunogen was the mouse preadipose cell line PA6.
Specifications
CD106 | |
Monoclonal | |
0.2mg/mL | |
Vcam-1; Vascular cell adhesion molecule 1; Vascular cell adhesion protein 1 | |
Rat | |
Affinity Purified | |
RUO | |
Mouse | |
IgG2a κ |
Flow Cytometry | |
429 (MVCAM.A) | |
Brilliant Violet 786 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse preadipose cell line PA6 | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only