Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD117 Rat anti-Mouse, Brilliant Violet 421, Clone: ACK45, BD Optibuild™

Rat Monoclonal Antibody

Manufacturer:  BD Biosciences 740036

Catalog No. BDB740036


Add to cart

Description

Description

The ACK45 monoclonal antibody specifically binds to CD117 (c-Kit), a transmembrane tyrosine-kinase receptor which is encoded by the Kit gene (formerly dominant white spotting, W ). The c-Kit ligand (also known as steel factor, stem cell factor, and mast-cell growth factor) encoded by the Kitl gene (formerly steel, Sl), is a co-mitogen for hematopoietic stem cells and the myeloerythroid progenitors and a mast-cell differentiation factor. The mutant alleles have similar pleiotropic effects on the development of melanocytes, germ cells, and the hematopoietic system. In the adult bone marrow, CD117 is expressed on hematopoietic progenitor cells, including the CD90 (Thy-1)[lo] TER-119[-] CD45R/B220[-] CD11b (Mac-1)[-] Ly-6G (Gr-1)[-] CD4[-] CD8[-] Sca-1 (Ly-6A/E)[+] multipotent hematopoietic stem cells, progenitors committed to myeloid and/or erythroid lineages, and precursors of B and T lymphocytes. This widespread expression of CD117 in hematopoietic precursors is consistent with the participation of c-Kit and its ligand in the regulation of several hematopoietic lineages. Intrathymic expression of c-Kit and c-Kit ligand suggest that CD117 is also involved in the regulation of some events during the development of T lymphocytes. CD117 is also expressed by mast cells. The expression of CD117 has been included in protocols to isolate hematopoietic progenitors from fetal and adult peripheral blood. The specificity and functional activity of mAb ACK45 is very similar to that of the published antibody ACK22; it is reported to block the action of c-Kit in vitro and in vivo.

Specifications

Specifications

CD117
ACK45
Brilliant Violet 421
Aqueous buffered solution containing ≤0.09% sodium azide.
Rat
IgG2b, κ
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
0.2mg/mL
c-KIT; W; SCFR; Stem Cell Factor Receptor; Sl; Steel Factor Receptor; Ssm
Affinity Purified
Mouse bone marrow mast cells
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.
RUO
Primary
Murine
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only