Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD122 Rat anti-Mouse, Brilliant Violet 711, Clone: 5H4, BD Optibuild™

Rat Monoclonal Antibody

Manufacturer:  BD Biosciences 740718

Catalog No. BDB740718


Add to cart

Description

Description

The 5H4 antibody monoclonal antibody specifically binds to mouse CD122. CD122 is a 70-85 kDa type I transmembrane glycoprotein of the hematopoietin receptor superfamily. CD122 is also known as IL-2 Receptor beta chain (IL-2R β ) or IL-15 Receptor beta chain (IL-15R β ) as it helps form signaling receptor complexes for Interleukin-2 (IL-2) or IL-15, respectively. CD122 can combine with either CD132 (γc) alone or CD132 plus CD25 (IL-2Rα) to form inte rmediate or high-affinity IL-2 Receptor complexes, respectively. The IL-2R β chain is constitutively expressed on NK cells and at lower levels on some thymocytes, T cells, B cells, monocytes, and macrophages. IL-2R β chain expression is up regulated by IL-2. The immunogen used to generate this hybridoma was rat myel oma cells transfected with a truncated IL-2R β cDNA.

The antibody was conjugated to BD Horizon™ BV711 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 711-nm. BD Horizon BV711 can be excited by the violet laser and detected in a filter used to detect Cy™5.5 / Alexa Fluor™ 700-like dyes (eg, 712/20-nm filter). Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy5.5 detectors. However, the spillover can be corrected through compensation as with any other dye combination.

Specifications

Specifications

CD122
5H4
Brilliant Violet 711
Aqueous buffered solution containing ≤0.09% sodium azide.
Rat
IgG2a, κ
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
0.2mg/mL
Il2rb; Il-2Rbeta; IL-2Rβ; IL-15Rbeta; IL-2/15 Receptor-beta; IL-2/15Rbeta
Affinity Purified
Rat myeloma cells transfected with a truncated IL-2R β cDNA
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.
RUO
Primary
Murine
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only