Learn More
CD125 Rat anti-Mouse, Brilliant Violet 650, Clone: T21, BD Optibuild™
Rat Monoclonal Antibody
Supplier: BD Biosciences 740623
Description
The T21 monoclonal antibody specifically binds to CD125 which is also known as the alpha subunit of the IL-5 Receptor (IL5RA/IL-5R α ). CD125 is a type I transmembrane protein that belongs to the hemopoietin receptor and Ig gene superfamilies. It is expressed on B cells and eosinophils. CD125 associates with the Common β chain ( β c/Bc) subunit, also known as CD131, to form the functional IL-5 Receptor complex. Mouse IL-5 belongs to the hematopoietic growth factor family of cytokines and promotes the growth and differentiation of B cells and eosinophils. Upon binding, the T21 antibody blocks IL-5 binding to its receptor and can inhibit IL-5 induced B-cell proliferation and antibody production.
Specifications
CD125 | |
Monoclonal | |
0.2mg/mL | |
Il5r; Il5ra; IL-5RA; IL-5Rα; IL-5R-alpha; IL-5R subunit alpha | |
Rat | |
Affinity Purified | |
RUO | |
Mouse | |
IgG1 λ |
Flow Cytometry | |
T21 | |
Brilliant Violet 650 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse T88-M Cell Line | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only