Learn More
CD126 Rat anti-Mouse, Brilliant Violet 421, Clone: D7715A7, BD Optibuild™
Rat Monoclonal Antibody
Supplier: BD Biosciences 740038
Description
The D7715A7 monoclonal antibody specifically binds to the mouse IL-6 receptor, which is also known as, CD126. The immunogen used to generate the D7715A7 hybridoma was OKT4 hybridoma cells. The D7715A7 hybridoma was selected based on its capacity to produce antibody that inhibited IL-6 binding to 2F4 cells (mouse B-cell hybridoma expressing high levels of IL-6 receptors). The binding of purified D7715A7 to B9 cells (mouse B-cell hybridoma expressing high levels of IL-6 receptors) is inhibited by recombinant mouse IL-6. This antibody has been reported to inhibit the in vitro and in vivo growth of the IL-6-dependent plasmacytoma line, T1033C2.
Specifications
CD126 | |
Monoclonal | |
0.2mg/mL | |
Il6ra; IL-6 Receptor α chain; IL-6RA; IL-6R alpha | |
Rat | |
Affinity Purified | |
RUO | |
Mouse | |
IgG2b κ |
Flow Cytometry | |
D7715A7 | |
Brilliant Violet 421 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
OKT4 hybridoma cells | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only