Learn More
CD13 Rat anti-Mouse, Brilliant Violet 510, Clone: R3-242, BD Optibuild™
Rat Monoclonal Antibody
Supplier: BD Biosciences 740127
Description
The R3-242 monoclonal antibody specifically binds to aminopeptidase N (APN), which has been reported to be the mouse CD13 homologue. APN is expressed in a variety of mammalian tissues, including intestinal and renal epithelial brush border, endothelium, liver, lung, and brain. In lymphoid tissues, R3-242 mAb is reported to detect APN on monocytes, macrophages, mast cells, dendritic cells, and B lymphocytes, but not on T cells, and thymocytes. However, the reported reactivity with B cells may be non-specific, via binding to the Fc γreceptors (CD16/CD32). This observation is in agreement with reports using different antibodies to mouse APN . APN may play a role in antigen presentation in the immune system. R3-242 mAb does not inhibit the enzymatic activity of APN.
The antibody was conjugated to BD Horizon™ BV510 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 405-nm and Em Max at 510-nm, BD Horizon BV510 can be excited by the violet laser and detected in the BD Horizon V500 (525/50-nm) filter set. BD Horizon BV510 conjugates are useful for the detection of dim markers off the violet laser.
Specifications
CD13 | |
Monoclonal | |
0.2mg/mL | |
Anpep; Alanyl aminopeptidase; Aminopeptidase N; AP-N; mAPN; Lap-1 | |
Rat | |
Affinity Purified | |
RUO | |
Mouse | |
IgG1 |
Flow Cytometry | |
R3-242 | |
Brilliant Violet 510 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Crude Aminopeptidase N Preparation from BALB/c Mouse Intestinal Epithelium | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only