Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD134 Rat anti-Mouse, Brilliant Violet 786, Clone: OX-86, BD Optibuild™

Rat Monoclonal Antibody

Manufacturer:  BD Biosciences 740945

Catalog No. BDB740945


Add to cart

Description

Description

The OX-86 monoclonal antibody specifically binds to the OX-40 antigen (CD134), also known as OX-40L receptor. CD134 is a 50-kDa type-I membrane glycoprotein that belongs to the NGFR/TNFR superfamily. Mouse CD134 is expressed on activated CD4+ and CD8+ T lymphocytes and has been shown to be the sole receptor for the OX-40 Ligand (OX-40L). In the brains of mice with actively induced experimental allergic encephalomyelitis, the expression of CD134 on CD4+ T lymphocytes correlates with disease progression. The OX-40/OX-40L system supplies a costimulatory signal for T-cell proliferation and B-cell proliferation and differentiation. In addition, OX-40 antigen provides a costimulatory signal that induces T cells to proliferate in a CD28-independent manner. In the intact animal, CD134 does not appear to be essential for many T-cell responses, but it seems to play a major role in the pathogenesis of some autoimmune diseases. The OX-86 mAb stains both CD4+ and CD8+ activated T cells, and this expression pattern has been confirmed using OX-40L-Ig fusion protein. CD134 was also detected, using OX-86 mAb, on B cells after stimulation with anti-IgM plus anti-CD40 mAb HM40-3 (Cat. no. 553721). OX-86 mAb does not block binding of OX-40L to OX-40, and it stimulates T-cell proliferation mildly.

The antibody was conjugated to BD Horizon™ BV786 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 786-nm. BD Horizon BV786 can be excited by the violet laser and detected in a filter used to detect Cy™7-like dyes (eg, 780/60-nm filter).

Specifications

Specifications

CD134
OX-86
Brilliant Violet 786
Aqueous buffered solution containing ≤0.09% sodium azide.
Rat
IgG1, κ
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
0.2mg/mL
Tnfrsf4; Ox40; OX40L receptor; Ly-70; ACT35; Txgp1; TXGP1L
Affinity Purified
Recombinant Mouse OX-40 3/4 CD4 Chimeric Protein
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.
RUO
Primary
Murine
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only