Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD137 Rat anti-Mouse, Brilliant Violet 711, Clone: 1AH2, BD Optibuild™

Rat Monoclonal Antibody

Manufacturer:  BD Biosciences 740707

Catalog No. BDB740707


Add to cart

Description

Description

The 1AH2 monoclonal antibody specifically binds to CD137. CD137 is a member of the TNFR/NGFR superfamily that is likewise known as Tnfrsf9, 4-1BB, Ly-63, or ILA. Monomers or multimeric forms of CD137 are expressed, upon activation, on the surface of splenic T lymphocytes, thymocytes, intestinal intraepithelial T lymphocytes (IEL), and some T cell lines and clones. While stimulating T cells by IL-2, IL-4, or anti-CD28 alone does not result in the expression of CD137; addition of IL-2, IL-4, anti-CD28, or syngeneic accessory cells to splenic T cells stimulated via TCR/CD3 can result in a high level CD137 expression. CD137 is also reportedly expressed on IL-2 activated NK cells, but not on freshly isolated NK cells. CD137 physically associates with p56 [lck] through a Cys-Arg-Cys-Pro binding site in its cytoplasmic domain; the same motif in the cytoplasmic tail of the CD4 and CD8a molecules is responsible for association with p56 [lck]. A signaling function for the CD137 molecule in mouse T cells is indicated by reports in which cross-linking of CD137 with 1AH2 mAb resulted in enhanced proliferation of CD3e-activated splenic T cells and IEL and in enhanced cytolytic activity of IEL in response to immobilized anti-CD3e. In addition to extracellular matrix proteins which bind to CD137, the CD137L (4-1BBL) serves as a ligand for CD137. This molecule has also been detected on LPS-activated macrophages, and anti-IgM antibody-activated splenic B cells. Interaction between T and B cells through CD137/CD137L is reported to play a role in antigen presentation, further supporting a costimulatory role for CD137 in the immune response of T lymphocytes.

The antibody was conjugated to BD Horizon™ BV711 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 711-nm. BD Horizon BV711 can be excited by the violet laser and detected in a filter used to detect Cy™5.5 / Alexa Fluor™ 700-like dyes (eg, 712/20-nm filter). Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy5.5 detectors. However, the spillover can be corrected through compensation as with any other dye combination.

Specifications

Specifications

CD137
1AH2
Brilliant Violet 711
Aqueous buffered solution containing ≤0.09% sodium azide.
Rat
IgG1, κ
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
0.2mg/mL
4-1BB; Tnfrsf9; Ly-63; ILA
Affinity Purified
Recombinant mouse 4-1BB
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.
RUO
Primary
Murine
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only