Learn More
CD140a Rat anti-Mouse, Brilliant Violet 605, Clone: APA5, BD Optibuild™
Rat Monoclonal Antibody
Supplier: BD Biosciences 740380
Description
The APA5 antibody monoclonal antibody specifically binds to CD140a, the Platelet-Derived Growth Factor Receptor alpha chain (PDGFR-a, PDGFR- α ). CD140a is a receptor tyrosine kinase that is widely expressed on cells of mesenchymal origin in the embryo and adult. CD140a is expressed on several other cell types during embryonic development, but not on hematopoietic cells. CD140a binds to PDGF A and B chains, in contrast to PDGFR-b (CD140b) that binds only to the PDGF B chain. Biologically active PDGF is a disulphide-linked dimer, forming the AA, AB, and BB isoforms. Ligand binding to the PDGF Receptor induces the formation of receptor dimers (aa, ab, or bb), autophosphorylation, and internalization. The APA5 antibody has been demonstrated to block binding of PDGF-AA to PDGFR-a-expressing cells in vitro and to block some PDGF-mediated developmental events in vivo.
This antibody is conjugated to BD Horizon™ BV605 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407-nm and Em Max of 602-nm, BD Horizon BV605 can be excited by a violet laser and detected with a standard 610/20-nm filter set. BD Horizon BV605 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an Em max at 605-nm. Due to the excitation of the acceptor dye by the green (532 nm) and yellow-green (561 nm) lasers, there will be significant spillover into the PE and BD Horizon PE-CF594 detectors off the green or yellow-green lasers. BD Horizon BV605 conjugates are very bright, often exhibiting brightness equivalent to PE conjugates and can be used as a third color off of the violet laser.
Specifications
CD140a | |
Monoclonal | |
0.2mg/mL | |
Pdgfra; Pdgfr2; PDGF-R alpha; Platelet derived growth factor receptor alpha | |
Rat | |
Affinity Purified | |
RUO | |
Mouse | |
IgG2a κ |
Flow Cytometry | |
APA5 | |
Brilliant Violet 605 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse PDGF Receptor α chain | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only