Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD18 Rat anti-Mouse, Brilliant Violet 650, Clone: C71/16, BD Optibuild™

Rat Monoclonal Antibody

Manufacturer:  BD Biosciences 740465

Catalog No. BDB740465


Add to cart

Description

Description

The C71/16 monoclonal antibody specifically binds to mouse CD18 which is also known as the common β 2 chain of LFA-1 (CD11a/CD18, α L β 2 integrin), Mac-1 (CD11b/CD18, α M β 2 integrin), and gp150, 95 (CD11c/CD18, α x β

The antibody was conjugated to BD Horizon™ BV650 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 650-nm. BD Horizon BV650 can be excited by the violet laser and detected in a filter used to detect APC-like dyes (eg, 660/20-nm filter). Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. However, the spillover can be corrected through compensation as with any other dye combination.

Specifications

Specifications

CD18
C71/16
Brilliant Violet 650
Aqueous buffered solution containing ≤0.09% sodium azide.
Rat
IgG2a, κ
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
0.2mg/mL
Itgb2; Integrin beta-2; ITB2; LAD; LCAMB; LFA-1/Mac-1/p150,95 subunit beta
Affinity Purified
Cell membrane glycoproteins from mouse T-cell lymphoma BW5147
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.
RUO
Primary
Murine
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only