Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD180 Rat anti-Mouse, Brilliant Violet 650, Clone: RP/14, BD Optibuild™

Rat Monoclonal Antibody

Manufacturer:  BD Biosciences 740558

Catalog No. BDB740558


Add to cart

Description

Description

The RP/14 monoclonal antibody specifically binds to CD180, the “radioprotective” 105 kDa (RP105) antigen expressed on mature B lymphocytes. Like other members of the Toll-like Receptor (TLR) family, the extracellular region of CD180 contains tandem repeats of a leucine-rich motif flanked by regions containing conserved cysteines. These characteristic structural components of TLR may mediate protein-protein interactions and regulation of signal transduction involved in innate immune responses. Furthermore, CD180 associates with the secretory protein MD-1 on the cell surface. Cross-linking of cell-surface CD180 with the RP/14 antibody induces activation signals in B lymphocytes. Despite the expression of CD180 on lymphocytes of all strains tested (BALB/c, BALB/c. xid , C3H, C57BL/6, CBA/N, and 129/Sv), some strains (BALB/c. xid , C57BL/6. xid , CBA/N, 129/Sv, and 129/Ola) display deficient responses to that stimulatory effects of mAb RP/14.

The antibody was conjugated to BD Horizon™ BV650 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 650-nm. BD Horizon BV650 can be excited by the violet laser and detected in a filter used to detect APC-like dyes (eg, 660/20-nm filter). Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. However, the spillover can be corrected through compensation as with any other dye combination.

Specifications

Specifications

CD180
RP/14
Brilliant Violet 650
Aqueous buffered solution containing ≤0.09% sodium azide.
Rat
IgG2a, κ
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
0.2mg/mL
Ly78; Ly-78; Lymphocyte antigen 78; RP105; Radioprotective 105 kDa protein
Affinity Purified
BALB/c mouse splenocytes
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.
RUO
Primary
Murine
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only