Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD2 Rat anti-Mouse, Brilliant Violet 605, Clone: RM2-5, BD Optibuild™

Rat Monoclonal Antibody

Manufacturer:  BD Biosciences 740338

Catalog No. BDB740338


Add to cart

Description

Description

The RM2-5 monoclonal antibody specifically binds to the immunoglobulin superfamily adhesion molecule, CD2 (LFA-2). CD2 is a type I transmembrane glycoprotein that serves as the the major receptor for CD48 in the mouse. CD2 is involved in T-cell activation, immunoregulation, and thymocyte maturation. In the mouse, CD2 is expressed on peripheral T lymphocytes, B lymphocytes, and NK cells, and a subpopulation of intraepithelial T lymphocytes. CD2 is expressed throughout mouse thymic ontogeny, except for distinct subsets of the CD4-CD8- early thymocytes. In the mouse bone marrow, CD2 is expressed on B220+ sIg+ CD43- pre-B cells, but not on CD43+ pro-B cells. The RM2-5 antibody is one of a set of five anti-mouse CD2 monoclonal antibodies that were classified into two groups according to their mutual competition in binding to cell surface CD2 and which block CD2-mediated cell-cell adhesion.

This antibody is conjugated to BD Horizon™ BV605 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407-nm and Em Max of 602-nm, BD Horizon BV605 can be excited by a violet laser and detected with a standard 610/20-nm filter set. BD Horizon BV605 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an Em max at 605-nm. Due to the excitation of the acceptor dye by the green (532 nm) and yellow-green (561 nm) lasers, there will be significant spillover into the PE and BD Horizon PE-CF594 detectors off the green or yellow-green lasers. BD Horizon BV605 conjugates are very bright, often exhibiting brightness equivalent to PE conjugates and can be used as a third color off of the violet laser.

Specifications

Specifications

CD2
RM2-5
Brilliant Violet 605
Aqueous buffered solution containing ≤0.09% sodium azide.
Rat
IgG2b, λ
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
0.2mg/mL
LFA-2; LFA-3 receptor; Ly37; Ly-37; Lymphocyte antigen 37; T11
Affinity Purified
Mouse BALB/c Thymocytes
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.
RUO
Primary
Murine
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only