Learn More
CD209a Rat anti-Mouse, Brilliant Violet 786, Clone: 5H10, BD Optibuild™
Rat Monoclonal Antibody
Manufacturer:  BD Biosciences 741023
Catalog No. BDB741023
Add to cart
Description
Description
The 5H10 monoclonal antibody specifically binds to mouse CD209a, or CIRE, which is the most closely related homolog of human DC-SIGN. In the mouse, several homologs of human CD209 (DC-SIGN) have been described. CD209a is a 238-amino acid type-II transmembrane C-type lectin, with an extracellular carbohydrate recognition domain. It can bind to mannosylated ligands in a Ca++ dependent manner. CD209a is expressed by some conventional CD4+ and CD4-CD8- dendritic cells (DC) but not by CD8+ DC. It is also expressed on monocytes, splenic plasmacytoid pre-DC, macrophages, and resident peritoneal cells. CD209a expression is downregulated upon cellular activation. CD209a is thought to play a role in the internalization and presentation of antigens to initiate immune responses.
The antibody was conjugated to BD Horizon™ BV786 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 786-nm. BD Horizon BV786 can be excited by the violet laser and detected in a filter used to detect Cy™7-like dyes (eg, 780/60-nm filter).
Specifications
Specifications
CD209a
5H10
Brilliant Violet 786
Aqueous buffered solution containing ≤0.09% sodium azide.
Rat
IgG2a, κ
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
0.2mg/mL
CD209; Cire; DC-SIGN; Dcsign; DC-SIGN1; CDSIGN
Affinity Purified
Peptide Derived from Extracellular Region of CD209a/CIRE
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.
RUO
Primary
Murine