Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD26 Rat anti-Mouse, Brilliant Violet 421, Clone: H194-112, BD Optibuild™

Rat Monoclonal Antibody

Manufacturer:  BD Biosciences 740021

Catalog No. BDB740021


Add to cart

Description

Description

The H194-112 monoclonal antibody specifically binds to CD26, which is also known as, Thymocyte-activating molecule (THAM), or dipeptidyl peptidase IV (DPP IV, Dpp4). CD26 is a ~220- kDa dimer formed of identical type-II transmembrane core polypeptides which undergo variable post-translational modifications. It is a multi-functional molecule with both ectopeptidase and signal-transducing activities. Studies with specific DPP IV inhibitors suggest that the enzymatic activity is involved in the mediation of T-cell activation events. The expression of CD26 is developmentally regulated in the thymus. Resting lymphoid cells of the bone marrow and peripheral B and T lymphocytes express low levels of CD26; bone-marrow and peritoneal myeloid cells do not. CD26 is also found on epithelial cells in the kidney, liver, small intestine, and lung. Cross-linked H194-112 mAb induces proliferation of immature and mature thymocytes in the presence of either IL-1 plus IL-2 or PMA; addition of IL-2 or IL-4 to PMA further enhances the activation.

The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407-nm and Em Max at 421-nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50-nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates.

Specifications

Specifications

CD26
H194-112
Brilliant Violet 421
Aqueous buffered solution containing ≤0.09% sodium azide.
Rat
IgG2a, κ
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
0.2mg/mL
Cd26; Dpp4; dipeptidylpeptidase 4; THAM, DPP IV
Affinity Purified
Cell-surface molecules solubilized from BALB/c mouse thymocytes
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.
RUO
Primary
Murine
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only