Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD282 Rat anti-Mouse, Brilliant Violet 421, Clone: 6C2, BD Optibuild™

Rat Monoclonal Antibody

Manufacturer:  BD Biosciences 740000

Catalog No. BDB740000


Add to cart

Description

Description

The 6C2 monoclonal antibody specifically binds to the mouse Toll like receptor 2 (TLR2) that is also known as CD282. CD282 is a type 1 transmembrane glycoprotein and a TLR family member. It is expressed on the surface of monocytes, macrophages, dentritic cells, granulocytes, endothelial and epithelial cells. This pattern recognition receptor is important for sensing and transducing signals in response to microbial cell wall components including lipoproteins, peptidoglycans and lipoteichoic acid. CD282 signals through intracellular MYD88 and TRAF6 leading to NF-kappa-B activation and the production of cytokines and inflammatory mediators by responding cells. TLR2 plays a major role in mediating inflammatory and innate immune responses.

The antibody was conjugated to BD Horizon™ BV421 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407-nm and Em Max at 421-nm, BD Horizon BV421 can be excited by the violet laser and detected in the standard Pacific Blue™ filter set (eg, 450/50-nm filter). BD Horizon BV421 conjugates are very bright, often exhibiting a 10 fold improvement in brightness compared to Pacific Blue conjugates.

Specifications

Specifications

CD282
6C2
Brilliant Violet 421
Aqueous buffered solution containing ≤0.09% sodium azide.
Rat
IgG2b, κ
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
0.2mg/mL
TLR2; Tlr2; Toll-like receptor 2; Ly105
Affinity Purified
Mouse TLR2 Transfected Cell Line
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.
RUO
Primary
Murine
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only