Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD31 Rat anti-Mouse, Brilliant Violet 510, Clone: 390, BD Optibuild™

Rat Monoclonal Antibody

Manufacturer:  BD Biosciences 740124

Catalog No. BDB740124


Add to cart

Description

Description

The 390 monoclonal antibody specifically binds to CD31, also known as PECAM-1 (platelet endothelial cell adhesion molecule). CD31 is a ~130 kDa integral membrane glycoprotein, a member of the immunoglobulin superfamily, that mediates homophilic and heterophilic cell-cell adhesion. CD31 is expressed constitutively on the surface of adult and embryonic endothelial cells and is weakly expressed on many peripheral leukocytes and platelets. It has also been detected on bone marrow-derived hematopoietic stem cells and embryonic stem cells. CD31 is involved in the transendothelial emigration of neutrophils, and neutrophil PECAM-1 appears to be down-regulated after extravasation into inflamed tissues. Multiple alternatively spliced isoforms are detected during early post-implantation embryonic development; this alternative splicing is involved in regulation of ligand specificity. CD38 and vitronectin receptor ( α v β 3 integrin, CD51/CD61) are proposed to be ligands for CD31. CD31-mediated endothelial cell-cell interactions are involved in angiogenesis. The 390 mAb inhibits a variety of in vitro and in vivo functions mediated by CD31.

The antibody was conjugated to BD Horizon™ BV510 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 405-nm and Em Max at 510-nm, BD Horizon BV510 can be excited by the violet laser and detected in the BD Horizon V500 (525/50-nm) filter set. BD Horizon BV510 conjugates are useful for the detection of dim markers off the violet laser.

Specifications

Specifications

CD31
390
Brilliant Violet 510
Aqueous buffered solution containing ≤0.09% sodium azide.
Rat
IgG2a, κ
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
0.2mg/mL
Platelet endothelial cell adhesion molecule; Pecam1; PECAM-1; Pecam-1
Affinity Purified
C3H/HeJ mouse hematopoietic progenitor cell line 32D
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.
RUO
Primary
Murine
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only