Learn More
CD38 Rat anti-Mouse, Brilliant Violet 786, Clone: 90/CD38 (also known as Ab90), BD Optibuild™
Rat Monoclonal Antibody
Supplier: BD Biosciences 740887
Description
The 90 monoclonal antibody specifically binds to CD38, a 42 kDa transmembrane glycoprotein on immature and mature, resting and activated, B lymphocytes. In contrast to humans, CD38 expression is down-regulated on mouse germinal center B cells and plasma cells. CD38 is also expressed on a subpopulation of thymic and peripheral T cells, NK cells, and splenic macrophages. Furthermore, CD38 has been detected on bone marrow-derived hematopoietic stem cells. The CD38 molecule is reported to exhibit both cyclase and hydrolase activities and plays a role in lymphocyte activation. CD31, both human and mouse, is reported to be a ligand for human CD38.
The antibody was conjugated to BD Horizon™ BV786 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 786-nm. BD Horizon BV786 can be excited by the violet laser and detected in a filter used to detect Cy™7-like dyes (eg, 780/60-nm filter).
Specifications
CD38 | |
Monoclonal | |
0.2mg/mL | |
ADP-ribosyl cyclase 1; Cyclic ADP-ribose hydrolase 1; I-19; NIM-R5 | |
Rat | |
Affinity Purified | |
RUO | |
Mouse | |
IgG2a κ |
Flow Cytometry | |
90/CD38 (also known as Ab90) | |
Brilliant Violet 786 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse Bone Marrow Pre-B cells | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only