Learn More
CD41 Rat anti-Mouse, Brilliant Violet 510, Clone: MWReg30, BD Optibuild™
Rat Monoclonal Antibody
Supplier: BD Biosciences 740136
Description
The MWReg30 monoclonal antibody specifically binds to CD41, which is also known as glycoprotein IIb (gpIIb, GP IIb), Integrin α IIb chain, or Integrin alpha 2b. CD41 is a transmembrane glycoprotein that is encoded by Itga2b . CD41 associates with I ntegrin β3 chain (g pIIIa or CD61) to form the gpIIb/IIIa (CD41/CD61) complex. CD41/CD61 is expressed on platelets, megakaryocytes, and early hematopoietic progenitors. The integrin complex binds to fibrinogen, fibronectin, vitronectin, von Willebrand factor, and thrombospondin. It is important for platelet adhesion and aggregation, and it may play a role in osteolytic tumor metastasis.
Specifications
CD41 | |
Monoclonal | |
0.2mg/mL | |
Itga2b; Integrin alpha 2b; Integrin alpha-IIb; Integrin α IIb; GpIIb | |
Rat | |
Affinity Purified | |
RUO | |
Mouse | |
IgG1 κ |
Flow Cytometry | |
MWReg30 | |
Brilliant Violet 510 | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse Platelets | |
50 μg | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only