Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD62E Rat anti-Mouse, Brilliant Violet 711, Clone: 10E9.6, BD Optibuild™

Rat Monoclonal Antibody

Manufacturer:  BD Biosciences 740694

Catalog No. BDB740694


Add to cart

Description

Description

The 10E9.6 monoclonal antibody specifically binds to the 97-110 kDa cell surface glycoprotein E-selectin (CD62E), also known as endothelial-leukocyte adhesion molecule-1 (ELAM-1), which is expressed on endotoxin- or cytokine-stimulated mouse endothelial cells. A suspension of TNF α stimulated mouse brain capillary endothelioma cells, from the cell line bEnd.3, was used as the immunogen. The epitope recognized by mAb 10E9.6 has been mapped to the first and/or second complement regulatory protein repeat domains of E-selectin. The 10E9.6 antibody has been reported to block binding of a monocyte cell line to E-selectin in vitro and to block neutrophil migration in BALB/c, but not C57BL/6 mice. It has no effect on leukocyte rolling in TNF α -treated mouse venules or on in vitro adhesion of myeloid cells to E-selectin. Studies have demonstrated that Cutaneous Lymphocyte Antigen (CLA), recognized by mAb HECA-452 (Cat. no. 555946), may be a ligand for CD62E.

The antibody was conjugated to BD Horizon™ BV711 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 711-nm. BD Horizon BV711 can be excited by the violet laser and detected in a filter used to detect Cy™5.5 / Alexa Fluor™ 700-like dyes (eg, 712/20-nm filter). Due to the excitation and emission characteristics of the acceptor dye, there may be moderate spillover into the Alexa Fluor™ 700 and PerCP-Cy5.5 detectors. However, the spillover can be corrected through compensation as with any other dye combination.

Specifications

Specifications

CD62E
10E9.6
Brilliant Violet 711
Aqueous buffered solution containing ≤0.09% sodium azide.
Rat
IgG2a, κ
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
0.2mg/mL
Sele; E-selectin; Elam; ELAM-1; LECAM2; LYAM2
Affinity Purified
Mouse brain capillary endothelioma bEnd.3 (TNFα-stimulated)
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.
RUO
Primary
Murine
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only