Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD74 Rat anti-Mouse, Brilliant Violet 605, Clone: In-1, BD Optibuild™

Rat Monoclonal Antibody

Manufacturer:  BD Biosciences 740385

Catalog No. BDB740385


Add to cart

Description

Description

The In-1 monoclonal antibody specifically binds to CD74, the MHC class II-associated Invariant chain (Ii), expressed in I-A+ cell populations. Although it is predominantly expressed intracellularly, it has also been detected on the cell surface of some B-cell lines and at low levels on a small number of B lymphocytes. In the mouse, alternatively spliced transcripts encode 31-kDa and 41-kDa isoforms, and the cell-surface CD74 has a chondroitin sulfate side chain which allows it to interact with CD44. CD74 is directly involved in the intracellular transport of MHC class II molecules and antigen presentation, and it affects the maturation of T and B lymphocytes.

This antibody is conjugated to BD Horizon™ BV605 which is part of the BD Horizon Brilliant™ Violet family of dyes. With an Ex Max of 407-nm and Em Max of 602-nm, BD Horizon BV605 can be excited by a violet laser and detected with a standard 610/20-nm filter set. BD Horizon BV605 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an Em max at 605-nm. Due to the excitation of the acceptor dye by the green (532 nm) and yellow-green (561 nm) lasers, there will be significant spillover into the PE and BD Horizon PE-CF594 detectors off the green or yellow-green lasers. BD Horizon BV605 conjugates are very bright, often exhibiting brightness equivalent to PE conjugates and can be used as a third color off of the violet laser.

Specifications

Specifications

CD74
In-1
Brilliant Violet 605
Aqueous buffered solution containing ≤0.09% sodium azide.
Rat
IgG2b, κ
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
0.2mg/mL
Cd74; CLIP; Ii; Ia-associated invariant chain; Ia-GAMMA; DHLAG; HG2A; HLADG
Affinity Purified
A.TL mouse lymphocytes and B10.A mouse-derived CH1 B-cell lymphoma
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.
RUO
Primary
Murine
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only