Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CD83 Rat anti-Mouse, Brilliant Violet 786, Clone: Michel-19, BD Optibuild™

Rat Monoclonal Antibody

Manufacturer:  BD Biosciences 741021

Catalog No. BDB741021


Add to cart

Description

Description

The Michel-19 antibody reacts with CD83, a member of the immunoglobulin superfamily that is expressed on mature dendritic cells and activated T lymphocytes. Furthermore, thymic cortical epithelial cells express Cd83 transcripts. CD83 is involved in the regulation of T-cell development and immune responses, and its ligand is found on a subpopulation of splenic B lymphocytes.

The antibody was conjugated to BD Horizon™ BV786 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 786-nm. BD Horizon BV786 can be excited by the violet laser and detected in a filter used to detect Cy™7-like dyes (eg, 780/60-nm filter).

Specifications

Specifications

CD83
Michel-19
Brilliant Violet 786
Affinity Purified
Mouse CD83 Recombinant Protein
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.
RUO
Primary
Murine
Flow Cytometry
0.2mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
Rat
IgG1, κ
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only