Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Early B Lineage Rat anti-Mouse, Brilliant Violet 786, Clone: 493, BD Optibuild™

Rat Monoclonal Antibody

Manufacturer:  BD Biosciences 740948

Catalog No. BDB740948


Add to cart

Description

Description

The 493 antibody reacts with a cell-surface protein of 130-140 kDa expressed on immature B lymphocytes and a small fraction of newly formed B cells, but not on mature B lymphocytes. The antigen’s distribution was defined through the use of antibodies to CD24 (Heat Stable Antigen), IgM, IgD, and CD45R/B220, which are commonly used to discriminate B-cell maturation stages. 493 mAb reacts with the majority of B220+ cells in bone marrow and a fraction of B220+ B cells in spleen, which are CD24[high], IgM[high], and IgD[low]. Cells binding 493 mAb were not detected in thymus, lymph nodes, or peritoneal cavity. This result suggested that 493 mAb does not stain B-1 B cells (CD5+ B lymphocytes), which are particularly found in the peritoneal cavity. The 493 mAb does not seem to have any biological effect when incubated with immature B lymphocytes. It has been observed that the staining pattern of mAb 493 is similar to that of mAb AA4.1, that both antibodies precipitate molecules of the same molecular weight, and that staining by mAb AA4.1 is not blocked by mAb 493, suggesting that the antibodies recognize separate epitopes of the same Early B Lineage antigen.

The antibody was conjugated to BD Horizon™ BV786 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405-nm and an acceptor dye with an Em Max at 786-nm. BD Horizon BV786 can be excited by the violet laser and detected in a filter used to detect Cy™7-like dyes (eg, 780/60-nm filter).

Specifications

Specifications

Early B Lineage
493
Brilliant Violet 786
Aqueous buffered solution containing ≤0.09% sodium azide.
Rat
IgG2a, κ
50μg
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Flow Cytometry
0.2mg/mL
Cd93; Ly68; C1qr1; C1qrp; Complement component C1q receptor
Affinity Purified
Pro-B cell line R2BFL derived from the fetal liver of a RAG-2-/-Bcl2-transgenic mouse
The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography.
RUO
Primary
Murine
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only